2-Dodecanone, 95%
2-Dodecanone, 95%
2-Dodecanone, 95%
2-Dodecanone, 95%
CAS: 6175-49-1 | C12H24O | 184.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA330290250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Dodecanone
CAS6175-49-1
CAS Min %94.0
CAS Max %100.0
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.