3-(2-Thienyl)propanoic acid, 98%
3-(2-Thienyl)propanoic acid, 98%
3-(2-Thienyl)propanoic acid, 98%
3-(2-Thienyl)propanoic acid, 98%
Thermo Scientific Chemicals
3-(2-Thienyl)propanoic acid, 98%
CAS: 5928-51-8 | C7H8O2S | 156.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA330110010
Price (MYR)
318.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
318.00
BOT
Add to cart
Specifications
Chemical Name or Material3-(2-Thienyl)propanoic acid
Name Note98%
CAS5928-51-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger