3-(2-Furyl)propanoic acid, 98%
3-(2-Furyl)propanoic acid, 98%
3-(2-Furyl)propanoic acid, 98%
3-(2-Furyl)propanoic acid, 98%
Thermo Scientific Chemicals
3-(2-Furyl)propanoic acid, 98%
CAS: 935-13-7 | C7H8O3 | 140.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA330100100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-(2-Furyl)propanoic acid
CAS935-13-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0