Citric acid, for molecular biology, anhydrous, Rnase and Protease free
Citric acid, for molecular biology, anhydrous, Rnase and Protease free
Citric acid, for molecular biology, anhydrous, Rnase and Protease free
Citric acid, for molecular biology, anhydrous, Rnase and Protease free
Thermo Scientific Chemicals
Citric acid, for molecular biology, anhydrous, Rnase and Protease free
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Catalog NumberQuantity
FSA3298600101 kg
Catalog number FSA329860010
Price (MYR)
537.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
537.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCitric acid
CAS77-92-9
Melting Point153.0°C
Health Hazard 1Warning
ColorWhite
View more
Biological buffer for molecular biology
RUO – Research Use Only