Heptadecanoic acid, 98%
Heptadecanoic acid, 98%
Heptadecanoic acid, 98%
Heptadecanoic acid, 98%
Thermo Scientific Chemicals
Heptadecanoic acid, 98%
CAS: 506-12-7 | C17H34O2 | 270.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA329740250
Price (MYR)
1,615.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,615.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHeptadecanoic acid
Name Note95%
CAS506-12-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: