n-Dodecyl-beta-D-maltoside, 99%, high purity
n-Dodecyl-beta-D-maltoside, 99%, high purity
n-Dodecyl-beta-D-maltoside, 99%, high purity
n-Dodecyl-beta-D-maltoside, 99%, high purity
Thermo Scientific Chemicals
n-Dodecyl-beta-D-maltoside, 99%, high purity
CAS: 69227-93-6 | C24H46O11 | 510.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA329370010
Price (MYR)
965.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
965.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Dodecyl-β-D-maltoside
CAS69227-93-6
CAS Min %98.5
CAS Max %100.0
Melting Point217.0°C