N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%
N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%
N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%
N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%
Thermo Scientific Chemicals
N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%
CAS: 51298-62-5 | C7H16ClN5O4 | 269.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA328750050
Price (MYR)
1,266.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,266.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-ω-Nitro-L-arginine-methyl ester hydrochloride
CAS51298-62-5
CAS Min %97.5
CAS Max %100.0