Daidzein, 98%
Daidzein, 98%
Daidzein, 98%
Thermo Scientific Chemicals
Daidzein, 98%
CAS: 486-66-8 | C15H10O4 | 254.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Packaging:
Glass bottle
Catalog number FSA328231000
Price (MYR)
1,623.00
EA
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,623.00
EA
Add to cart
Specifications
Chemical Name or MaterialDaidzein
CAS486-66-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a