AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
Thermo Scientific Chemicals
AEBSF hydrochloride, 98%
CAS: 30827-99-7 | C8H10FNO2S·ClH | 239.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
1 g
Packaging:
Glass bottle
Catalog number FSA328110500
Price (MYR)
411.00
BOT
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
411.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAEBSF hydrochloride
Name Note98%
CAS30827-99-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger