Methanol, 99.8%, for electronic use (MOS), residue free
Methanol, 99.8%, for electronic use (MOS), residue free
Methanol, 99.8%, for electronic use (MOS), residue free
Methanol, 99.8%, for electronic use (MOS), residue free
Thermo Scientific Chemicals
Methanol, 99.8%, for electronic use (MOS), residue free
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA327900025
Price (MYR)
482.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
482.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
Name NoteResidue Free
CAS67-56-1
Health Hazard 1Danger
View more
For electronic use
RUO – Research Use Only