Tricine, 99+%, for biochemistry
Tricine, 99+%, for biochemistry
Tricine, 99+%, for biochemistry
Tricine, 99+%, for biochemistry
Thermo Scientific Chemicals
Tricine, 99+%, for biochemistry
CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA327831000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance0.02 max. at 280nm
Chemical Name or MaterialTricine
Absorbance Conditions(0.1M aq. soln.)
CAS5704-04-1
Melting Point180.0°C to 184.0°C