TAPSO, for biochemistry
TAPSO, for biochemistry
TAPSO, for biochemistry
TAPSO, for biochemistry
Thermo Scientific Chemicals
TAPSO, for biochemistry
CAS: 68399-81-5 | C7H17NO7S | 259.273 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA327811000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Absorbance(0.1 M aq. soln.)
Chemical Name or MaterialTAPSO
Absorbance Conditions(0.1M aq. soln.)
CAS68399-81-5
Melting Point220.0°C to 221.0°C
View more
Biological buffer
RUO – Research Use Only