MES monohydrate, 99+%, for biochemistry
MES monohydrate, 99+%, for biochemistry
MES monohydrate, 99+%, for biochemistry
MES monohydrate, 99+%, for biochemistry
Thermo Scientific Chemicals
MES monohydrate, 99+%, for biochemistry
CAS: 145224-94-8 | C6H15NO5S | 195.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA327765000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance(0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Chemical Name or MaterialMES monohydrate
Absorbance Conditions(0.1M aq. soln.)
CAS145224-94-8
Melting Point>300.0°C
View more
Biological buffer
RUO – Research Use Only