Bicine, 99+%, for biochemistry
Bicine, 99+%, for biochemistry
Bicine, 99+%, for biochemistry
Bicine, 99+%, for biochemistry
Thermo Scientific Chemicals
Bicine, 99+%, for biochemistry
CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA327711000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance0.05 max. (0.5M, water) at 260nm, 0.05 max. (0.5M, water) at 280nm
Chemical Name or MaterialBicine
Name NoteFor biochemistry
CAS150-25-4
Melting Point190.0°C to 192.0°C
View more
Biological buffer
RUO – Research Use Only