4-Methoxychalcone, 98%, Thermo Scientific™
4-Methoxychalcone, 98%, Thermo Scientific™
4-Methoxychalcone, 98%, Thermo Scientific™
4-Methoxychalcone, 98%, Thermo Scientific™
4-Methoxychalcone, 98%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA327640250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Methoxychalcone
CAS959-33-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0