Water, for molecular biology, DNAse, RNAse and Protease free
Water, for molecular biology, DNAse, RNAse and Protease free
Water, for molecular biology, DNAse, RNAse and Protease free
Water, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals
Water, for molecular biology, DNAse, RNAse and Protease free
Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, 100.0 deg.C, CHEBI:15377, 0.1ppm max., Undesignated, 1.0000g/mL, none detected, 16,383, 18.02, Molecular Biology, XLYOFNOQVPJJNP-UHFFFAOYSA-N, oxidane, 1ppb max., 15, 10237, MFCD00011332, 18.015, DNase, RNase, Protease free | CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
View all
Plastic Drum
Catalog number FSA327390050
Price (MYR)
1,351.00
BOT
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
1,351.00
BOT
Add to cart
Specifications
Chemical Name or MaterialWater
Name NoteDNase, RNase, Protease free
CAS7732-18-5
Health Hazard 1
Boiling Point100.0°C
View more
Biological buffer
RUO – Research Use Only