Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals
Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA327300010
Price (MYR)
513.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
513.00
BOT
Add to cart
Specifications
Boiling Point1461.0°C
Chemical Name or MaterialSodium chloride
Melting Point801.0°C
Name NoteDNAse, RNAse and Protease Free
CAS7647-14-5
View more
Biological buffer
RUO – Research Use Only