5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, RNAse and Protease free
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, RNAse and Protease free
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, RNAse and Protease free
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, RNAse and Protease free
CAS: 7240-90-6 | C14H15BrClNO6 | 408.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Packaging:
Glass bottle
Catalog number FSA327245000
Price (MYR)
1,032.00
EA
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,032.00
EA
Add to cart
Specifications
Chemical Name or Material5-Bromo-4-chloro-3-indolyl-β-D-galactoside
Name NoteFor molecular biology
CAS7240-90-6
CAS Min %99.0
CAS Max %100.0