Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for molecular biology, DNAse, RNAse and Protease free
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for molecular biology, DNAse, RNAse and Protease free
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for molecular biology, DNAse, RNAse and Protease free
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for molecular biology, DNAse, RNAse and Protease free
CAS: 6381-92-6 | C10H18N2Na2O10 | 372.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA327205000
Price (MYR)
1,372.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,372.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylenediaminetetraacetic acid, disodium salt dihydrate
CAS6381-92-6
Absorbance(0.1M in water), (0.1 M in water) at 0nm, 0.02 max. at 280nm, 0.2 max. at 260nm
Health Hazard 1Acute toxicity (category 4), Specific target organ toxicity after repeated exposure (category 2)
Health Hazard 2Harmful if inhaled, May cause damage to organs through prolonged or repeated exposure if inhaled
View more
Chelation/Complexation reagent for molecular biology
RUO – Research Use Only