DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free
DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free
DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free
DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals
DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free
CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3271900101 gGlass Bottle
Catalog number FSA327190010
Price (MYR)
380.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
380.00
EA
Add to cart
Specifications
Chemical Name or MaterialDL-1, 4-Dithiothreitol
Name NoteDNase, RNase, Protease free
CAS3483-12-3
Absorbance(0.1 M in water), (0.1 M in water) at 0nm, 0.1 max. at 280nm, 0.5 max. at 260nm
Health Hazard 1GHS Signal Word: Warning