N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA327171000
Price (MYR)
353.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
353.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide
Melting Point-61.0°C
Name NoteDNase, RNase, Protease Free
Absorbance(undiluted), (undiluted) at 0nm, 0.1 max. at 280nm, 1 max. at 270nm
CAS68-12-2
View more
For molecular biology
RUO – Research Use Only