Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
Thermo Scientific Chemicals
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA326970025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrahydrofuran
Melting Point-108.4°C
Name NoteStabilized, water < 50ppm
CAS109-99-9