Methanol, 99.9%, Extra Dry, AcroSeal™
Methanol, 99.9%, Extra Dry, AcroSeal™
Methanol, 99.9%, Extra Dry, AcroSeal™
Methanol, 99.9%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Methanol, 99.9%, Extra Dry, AcroSeal™
Methanol, 99.9%, CH4O, CAS Number-67-56-1, methylol, methyl alcohol, colonial spirit, carbinol, wood alcohol, wood naphtha, pyroxylic spirit, methyl hydroxide, wood spirit, columbian spirit, 1L, 0.002% max. (as HCOOH), 0.0005 meq/g max., 64.7 deg.C, CHEBI:17790, Colorless, 0.7910g/mL, 200-659-6 | CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA326950025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
CAS67-56-1
ColorColorless
Health Hazard 1Danger