Ethyl acetate, 99.9%, Extra Dry, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA326900010
Price (MYR)
614.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
614.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-83.5°C
Name NoteWater <50ppm
CAS141-78-6