1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
Thermo Scientific Chemicals
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 L
Catalog number FSA326891000
Price (MYR)
416.00
BOT
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
416.00
BOT
Add to cart
Specifications
Chemical Name or Material1,4-Dioxane
Melting Point12.0°C
Name NoteWater <50ppm
CAS123-91-1
CAS Min %99.75