Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA326820025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteWater <50 ppm, Stabilized
CAS67-66-3
Health Hazard 1Danger