n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
Thermo Scientific Chemicals
n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA326780025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Hexane
Melting Point-95.0°C
Absorbance0.004 max. at 270nm, 0.097 max. at 220nm, 0.602 max. at 200nm
CAS110-54-3
CAS Min %94.0
View more
For residue analysis
RUO – Research Use Only