n-Heptane, 99+%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
n-Heptane, 99+%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
n-Heptane, 99+%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
n-Heptane, 99+%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, for trace analysis of polyaromatic hydrocarbons
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA326770025
Price (MYR)
1,102.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,102.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Heptane
Melting Point-91.0°C
Name NoteTested for Trace Analysis of Polyaromatic Hydrocarbons
CAS142-82-5
CAS Min %99.0
View more
For residue analysis
RUO – Research Use Only