Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
Thermo Scientific Chemicals
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number FSA326720025
Price (MYR)
676.00
LT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
676.00
LT
Add to cart
Specifications
Chemical Name or MaterialPetroleum ether
Name NoteECD Tested
CAS64742-49-0
Health Hazard 1Danger
Health Hazard 2GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
View more
For residue analysis
RUO – Research Use Only