Acetonitrile, 99.9%, for residue analysis, ECD tested
Acetonitrile, 99.9%, for residue analysis, ECD tested
Acetonitrile, 99.9%, for residue analysis, ECD tested
Acetonitrile, 99.9%, for residue analysis, ECD tested
Thermo Scientific Chemicals
Acetonitrile, 99.9%, for residue analysis, ECD tested
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA326680025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetonitrile
Melting Point-46.0°C
Name NoteECD tested, For pesticide analysis
CAS75-05-8
CAS Min %99.85
View more
For residue analysis
RUO – Research Use Only