Chloroform, 99.9%, for residue analysis, ECD tested for pesticide anal., stab. with Amylene
Chloroform, 99.9%, for residue analysis, ECD tested for pesticide anal., stab. with Amylene
Chloroform, 99.9%, for residue analysis, ECD tested for pesticide anal., stab. with Amylene
Chloroform, 99.9%, for residue analysis, ECD tested for pesticide anal., stab. with Amylene
Thermo Scientific Chemicals
Chloroform, 99.9%, for residue analysis, ECD tested for pesticide anal., stab. with Amylene
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3266700252.5 LGlass Bottle
Catalog number FSA326670025
Price (MYR)
766.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
766.00
EA
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteECD Tested, For Pesticide Analysis, Residue <0.0005%
CAS67-66-3
Health Hazard 1Warning
View more
For residue analysis
RUO – Research Use Only