n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane
n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane
n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane
n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane
Thermo Scientific Chemicals
n-Hexane, for pesticide residue analysis, approx. 95% n-Hexane
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA326660025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Hexane
Melting Point-95.0°C
Name NoteECD Tested, for Pesticide Analysis, Residue <0.0003%
CAS110-54-3
Health Hazard 1Danger
View more
For residue analysis
RUO – Research Use Only