Ethyl acetate, for pesticide residue analysis
Ethyl acetate, for pesticide residue analysis
Ethyl acetate, for pesticide residue analysis
Ethyl acetate, for pesticide residue analysis
Thermo Scientific Chemicals
Ethyl acetate, for pesticide residue analysis
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA326610025
Price (MYR)
475.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
475.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-83.5°C
Name NoteECD Tested, For Pesticide Analysis
CAS141-78-6
ColorColorless
View more
For residue analysis
RUO – Research Use Only