Cyclohexane, for pesticide residue analysis
Cyclohexane, for pesticide residue analysis
Cyclohexane, for pesticide residue analysis
Cyclohexane, for pesticide residue analysis
Thermo Scientific Chemicals
Cyclohexane, for pesticide residue analysis
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA326590010
Price (MYR)
447.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
447.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCyclohexane
Melting Point6.5°C
Name NoteECD Tested, For Pesticide Analysis
CAS110-82-7
Health Hazard 1Danger
View more
For residue analysis
RUO – Research Use Only