3-Bromopropionic acid, 97%
3-Bromopropionic acid, 97%
3-Bromopropionic acid, 97%
3-Bromopropionic acid, 97%
Thermo Scientific Chemicals
3-Bromopropionic acid, 97%
CAS: 590-92-1 | C3H5BrO2 | 152.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA325775000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Bromopropionic acid
CAS590-92-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0