Dichloromethane-d2, for NMR, 99.8 atom% D
Dichloromethane-d2, for NMR, 99.8 atom% D
Dichloromethane-d2, for NMR, 99.8 atom% D
Dichloromethane-d2, for NMR, 99.8 atom% D
Thermo Scientific Chemicals
Dichloromethane-d2, for NMR, 99.8 atom% D
CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Packaging:
Glass Bottle
Catalog number FSA321270100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDichloromethane-d2
Density1.3600g/mL
Deuterium (D)99.75% min.
PackagingGlass Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only