N,O-Bis(trimethylsilyl)trifluoroacetamide, 98%, packaged in 1 ml ampoules
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98%, packaged in 1 ml ampoules
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98%, packaged in 1 ml ampoules
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98%, packaged in 1 ml ampoules
Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98%, packaged in 1 ml ampoules
CAS: 25561-30-2 | C8H18F3NOSi2 | 257.39 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA32120010010 mLGlass Bottle
Catalog number FSA321200100
Price (MYR)
975.00
BOT
Add to cart
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
975.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, O-Bis(trimethylsilyl)trifluoroacetamide
Name Note98%
CAS25561-30-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
View more
Silylation reagent
RUO – Research Use Only