3,4-Dichlorobenzyl isocyanate, 98%
3,4-Dichlorobenzyl isocyanate, 98%
3,4-Dichlorobenzyl isocyanate, 98%
3,4-Dichlorobenzyl isocyanate, 98%
Thermo Scientific Chemicals
3,4-Dichlorobenzyl isocyanate, 98%
CAS: 19752-09-1 | C8H5Cl2NO | 202.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA319220100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 4-Dichlorobenzyl isocyanate
CAS19752-09-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0