Osmium tetroxide, 4 wt.% solution in water
Osmium tetroxide, 4 wt.% solution in water
Osmium tetroxide, 4 wt.% solution in water
Osmium tetroxide, 4 wt.% solution in water
Thermo Scientific Chemicals
Osmium tetroxide, 4 wt.% solution in water
CAS: 20816-12-0 | O4Os | 254.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA319010100
Price (MYR)
1,192.00
EA
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,192.00
EA
Add to cart
Chemical Identifiers
CAS20816-12-0, 7732-18-5
Specifications
Boiling Point100.0°C
Chemical Name or MaterialOsmium tetroxide
CAS20816-12-0
ColorYellow
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only