Cacodylic acid, 98%
Cacodylic acid, 98%
Cacodylic acid, 98%
Thermo Scientific Chemicals
Cacodylic acid, 98%
CAS: 75-60-5 | C2H7AsO2 | 138.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Packaging:
Glass bottle
Catalog number FSA318150100
Price (MYR)
492.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
492.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCacodylic acid
CAS75-60-5
Melting Point195°C to 198°C
Health Hazard 1GHS Signal Word: Danger