Bismuth, 99%, powder, -100 mesh, Thermo Scientific Chemicals
Bismuth, 99%, powder, -100 mesh, Thermo Scientific Chemicals
Bismuth, 99%, powder, -100 mesh, Thermo Scientific Chemicals
Bismuth, 99%, powder, -100 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bismuth, 99%, powder, -100 mesh, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number FSA318095000
Price (MYR)
1,687.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,687.00
EA
Add to cart
Specifications
Chemical Name or MaterialBismuth
ColorBlack
Name Note99.5%
CAS7440-69-9
Melting Point271.0°C