Gallium, 99.99%, (trace metal basis)
Gallium, 99.99%, (trace metal basis)
Gallium, 99.99%, (trace metal basis)
Gallium, 99.99%, (trace metal basis)
Thermo Scientific Chemicals
Gallium, 99.99%, (trace metal basis)
CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Plastic bottle
Catalog number FSA317990500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialGallium
ColorGray
CAS7440-55-3
Melting Point29.7°C
Boiling Point2400.0°C
View more
For ultra trace analysis
RUO – Research Use Only