Tin, 99.5%, powder, -100 mesh
Tin, 99.5%, powder, -100 mesh
Tin, 99.5%, powder, -100 mesh
Tin, 99.5%, powder, -100 mesh
Thermo Scientific Chemicals
Tin, 99.5%, powder, -100 mesh
CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA317811000
Price (MYR)
318.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
318.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTin
ColorGray
Name Note-100 mesh
CAS7440-31-5
Melting Point231.9°C
View more
Reduction reagent
RUO – Research Use Only