Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
CAS: 19598-90-4 | GdN3O9·6H2O | 451.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA316761000
Price (MYR)
1,437.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,437.00
EA
Add to cart
Specifications
Chemical Name or MaterialGadolinium(III) nitrate hexahydrate
Melting Point91°C
CAS19598-90-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
for ultra trace analysis
RUO – Research Use Only