Rhenium pentacarbonyl chloride, 98%
Rhenium pentacarbonyl chloride, 98%
Rhenium pentacarbonyl chloride, 98%
Rhenium pentacarbonyl chloride, 98%
Thermo Scientific Chemicals
Rhenium pentacarbonyl chloride, 98%
CAS: 14099-01-5 | C5ClO5Re | 361.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA316310050
Price (MYR)
6,116.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,116.00
BOT
Add to cart
Specifications
Chemical Name or MaterialRhenium pentacarbonyl chloride
Name Note98%
CAS14099-01-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: