Gadolinium(III) oxide, 99.9%, (trace metal basis)
Gadolinium(III) oxide, 99.9%, (trace metal basis)
Gadolinium(III) oxide, 99.9%, (trace metal basis)
Gadolinium(III) oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Gadolinium(III) oxide, 99.9%, (trace metal basis)
CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA315511000
Price (MYR)
1,486.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,486.00
EA
Add to cart
Specifications
Chemical Name or MaterialGadolinium(III) oxide
Melting Point1900.0°C
CAS12064-62-9
ColorWhite
Health Hazard 1GHS Signal Word: Warning
View more
for ultra trace analysis
RUO – Research Use Only