Boric acid, 99.99%, (trace metal basis), extra pure
Boric acid, 99.99%, (trace metal basis), extra pure
Boric acid, 99.99%, (trace metal basis), extra pure
Boric acid, 99.99%, (trace metal basis), extra pure
Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA315185000
Price (MYR)
823.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
823.00
EA
Add to cart
Specifications
Chemical Name or MaterialBoric acid
ColorWhite
Name NoteExtra Pure
CAS10043-35-3
Melting Point169.0°C
View more
For ultra trace analysis, Lewis acid
RUO – Research Use Only