Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous
Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous
Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous
Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous
Thermo Scientific Chemicals
Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous
CAS: 10024-93-8 | Cl3Nd | 250.59 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3150700505 gGlass bottle
Catalog number FSA315070050
Price (MYR)
790.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
790.00
EA
Add to cart
Specifications
Chemical Name or MaterialNeodymium(III) chloride
Melting Point784°C
Name Note99.9%
CAS10024-93-8
ColorWhite
View more
for ultra trace analysis
RUO – Research Use Only