N,N'-Dimethyl-1,3-propanediamine, 97%
N,N'-Dimethyl-1,3-propanediamine, 97%
N,N'-Dimethyl-1,3-propanediamine, 97%
N,N'-Dimethyl-1,3-propanediamine, 97%
Thermo Scientific Chemicals
N,N'-Dimethyl-1,3-propanediamine, 97%
CAS: 111-33-1 | C5H16N2 | 104.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA314830050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N′-Dimethyl-1,3-propanediamine
Name Note97%
CAS111-33-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger