Tris(2-aminoethyl)amine, 98+%
Tris(2-aminoethyl)amine, 98+%
Tris(2-aminoethyl)amine, 98+%
Tris(2-aminoethyl)amine, 98+%
Thermo Scientific Chemicals
Tris(2-aminoethyl)amine, 98+%
CAS: 4097-89-6 | C6H18N4 | 146.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA314791000
Price (MYR)
1,306.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,306.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTris(2-aminoethyl)amine
CAS4097-89-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth